Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis(diphenylamino)anthraquinone 96.0+%, TCI America™
SDP

Catalog No. B50921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50921G 1 g
B5092200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50921G Supplier TCI America Supplier No. B50921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 868850-50-4
Molecular Formula C38H26N2O2
Molecular Weight (g/mol) 542.64
MDL Number MFCD20040460
InChI Key TZENEWLXCXPNFX-UHFFFAOYSA-N
PubChem CID 58927077
IUPAC Name 2,6-bis(diphenylamino)-9,10-dihydroanthracene-9,10-dione
SMILES O=C1C2=C(C=C(C=C2)N(C2=CC=CC=C2)C2=CC=CC=C2)C(=O)C2=C1C=C(C=C2)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 542.64
Percent Purity ≥96.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(diphenylamino)anthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.