Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene 99.0+%, TCI America™
SDP

Catalog No. D4568200MG
Click to view available options
Quantity:
200 mg

Chemical Identifiers

CAS 365547-21-3
Molecular Formula C25H36Br2S2
Molecular Weight (g/mol) 560.491
MDL Number MFCD16619302
InChI Key NEQRAFHWWLGKEJ-UHFFFAOYSA-N
PubChem CID 68168413
SMILES CCCCC(CC)CC1(C2=C(C3=C1C=C(S3)Br)SC(=C2)Br)CC(CC)CCCC

Specifications

Color Green-Yellow
Quantity 200 mg
Formula Weight 560.49
Percent Purity ≥99.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b′]dithiophene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.