Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromo-4-(trifluoromethoxy)aniline 98.0+%, TCI America™
SDP

Catalog No. D303825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D303825G 25 g
D30385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D303825G Supplier TCI America Supplier No. D303825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88149-49-9
Molecular Formula C7H4Br2F3NO
Molecular Weight (g/mol) 334.92
MDL Number MFCD00153113
InChI Key JBSWOEGXMADXOU-UHFFFAOYSA-N
Synonym 2,6-dibromo-4-trifluoromethoxy aniline, bromo riluzole impurity, 3,5-dibromo-4-aminotrifluoromethoxybenzene, 2,6-dibromo-4-trifluoromethoxy benzenamine, 2-amino-1,3-dibromo-5-trifluoromethoxy benzene, 2,6dibromo-4-trifluoromethoxy aniline, 2,6-dibromo-4-trifluoro-methoxy aniline, benzenamine, 2,6-dibromo-4-trifluoromethoxy, pubchem10031
PubChem CID 2736797
IUPAC Name 2,6-dibromo-4-(trifluoromethoxy)aniline
SMILES NC1=C(Br)C=C(OC(F)(F)F)C=C1Br

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 334.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dibromo-4-(trifluoromethoxy)aniline
EINECSNumber (3)-4290
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.