Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromoaniline, 97%

Catalog No. p-7043554
Click to view available options
Quantity:
2 g
10 g

CAS: 608-30-0 | C6H5Br2N | 250.92 g/mol

2,5-Dibromoaniline was used in the synthesis of 2,4,5-tribromoaniline1. It was also used as starting reagent in the regioselective synthesis of 2-(3,4-dimethoxyphenyl)-5-bromobenzothiazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromoaniline was used in the synthesis of 2,4,5-tribromoaniline1. It was also used as starting reagent in the regioselective synthesis of 2-(3,4-dimethoxyphenyl)-5-bromobenzothiazole.

Solubility
Insoluble in water.

Notes
Incompatible with acids, Acid chlorides, Acid anhydrides, Oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-30-0
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.92
MDL Number MFCD00007638
InChI Key XIRRDAWDNHRRLB-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dibromo, 2,6-dibromophenylamine, 2,6-dibromobenzenamine, aniline, 2,6-dibromo, 2,6-dibromo-phenylamine, 2,6-dibromo-aniline, acmc-1aycf, 2,6-dibromoaniline, 2,6-dibromo-phenyl-amine, ksc356c3l
PubChem CID 69098
IUPAC Name 2,6-dibromoaniline
SMILES NC1=C(Br)C=CC=C1Br

Specifications

Melting Point 79°C to 83°C
Boiling Point 262°C to 264°C
Odor Characteristic
Quantity 2 g
UN Number UN2811
Beilstein 2207094
Solubility Information Insoluble in water.
Formula Weight 250.93
Percent Purity 97%
Chemical Name or Material 2,6-Dibromoaniline
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-158-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.