Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dibromonaphthalene, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13720-06-4
C10H6Br2
285.97
PJZDEYKZSZWFPX-UHFFFAOYSA-N
naphthalene, 2,6-dibromo,2,6-dibromo-naphthalene,2,6-dibromo naphthalene,2.6-dibromonaphthalene,2,6-?dibromonaphthalene,acmc-1c7c1,ksc490o3h,2,6-dibromonaphthalene
640591
2,6-dibromonaphthalene
C1=CC2=C(C=CC(=C2)Br)C=C1Br
13720-06-4
C10H6Br2
285.97
PJZDEYKZSZWFPX-UHFFFAOYSA-N
naphthalene, 2,6-dibromo,2,6-dibromo-naphthalene,2,6-dibromo naphthalene,2.6-dibromonaphthalene,2,6-?dibromonaphthalene,acmc-1c7c1,ksc490o3h,2,6-dibromonaphthalene
640591
2,6-dibromonaphthalene
C1=CC2=C(C=CC(=C2)Br)C=C1Br
Melting Point 162°C to 167°C
Packaging Glass bottle
Quantity 25g
Assay Percent Range 98.5% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 285.97
Percent Purity 99%
Chemical Name or Material 2, 6-Dibromonaphthalene