Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-1,4-benzoquinone, 97+%

Catalog No. AAH2754206
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH2754206

Supplier: Thermo Scientific Chemicals H2754206

Only null left
Add to Cart

CAS: 697-91-6 | C6H2Cl2O2 | 176.98 g/mol

2,6-Dichloro-1,4-benzoquinone is the suitable reagent used to in a study to evaluate the diffusion coefficients (D) for a family of quinones, nitroaromatics, ferrocenes and aromatic hydrocarbon compounds, in acetonitrile by single potential step chronoamperometry.1 It may be used in the preparation of 2,3,5-Trichloro-1,4-dihydroquinone, 2,3,5-trichloro-1,4-benzoquinone and 2,6-Dichloro-1,4-dihydroquinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloro-1,4-benzoquinone is the suitable reagent used to in a study to evaluate the diffusion coefficients (D) for a family of quinones, nitroaromatics, ferrocenes and aromatic hydrocarbon compounds, in acetonitrile by single potential step chronoamperometry.1 It may be used in the preparation of 2,3,5-Trichloro-1,4-dihydroquinone, 2,3,5-trichloro-1,4-benzoquinone and 2,6-Dichloro-1,4-dihydroquinone.

Solubility
Insoluble in water. Solubility in methanol (almost transparency).

Notes
Store away from reducing agents and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-91-6
Molecular Formula C6H2Cl2O2
Molecular Weight (g/mol) 176.98
MDL Number MFCD00037159
InChI Key JCARTGJGWCGSSU-UHFFFAOYSA-N
Synonym 2,6-dichloro-1,4-benzoquinone, 2,6-dichloro-p-benzoquinone, 2,6-dichlorobenzoquinone, 2,6-dichloroquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dichloro, 2,6-dichloquinone, p-quinone, 2,6-dichloro, p-benzoquinone, 2,6-dichloro, 2,6-dichloro-2,5-cyclohexadiene-1,4-dione, p-quinone,6-dichloro
PubChem CID 12771
IUPAC Name 2,6-dichlorocyclohexa-2,5-diene-1,4-dione
SMILES C1=C(C(=O)C(=CC1=O)Cl)Cl

Specifications

Melting Point 119°C to 121°C
Quantity 5 g
Solubility Information Insoluble in water. Solubility in methanol (almost transparency).
Formula Weight 176.98
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-1,4-benzoquinone
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 211-810-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.