Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-3-fluoroacetophenone, 97%, Thermo Scientific™

Catalog No. p-105137
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 290835-85-7
Molecular Formula C8H5Cl2FO
Molecular Weight (g/mol) 207.03
MDL Number MFCD02093760
InChI Key VJBFZHHRVCPAPZ-UHFFFAOYSA-N
Synonym 2',6'-dichloro-3'-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethanone, 2,6-dichloro-3-fluoroacetophenone, 1-2,6-dichloro-3-fluorophenyl ethan-1-one, 2,6-dichloro-3-fluro acetophenone, 2,6-dichloro-5-fluoro acetophenone, 2',6'-dichloro-3'-fluroacetophenone, ethanone, 1-2,6-dichloro-3-fluorophenyl, 1-acetyl-2,6-dichloro-3-fluorobenzene, pubchem8532
PubChem CID 2733982
IUPAC Name 1-(2,6-dichloro-3-fluorophenyl)ethanone
SMILES CC(=O)C1=C(Cl)C=CC(F)=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 250.0°C to 255.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5275 to 1.5295
Quantity 100 g
Formula Weight 207.03
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 6-Dichloro-3-fluoroacetophenone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.