Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-p-cresol 96.0+%, TCI America™
SDP

Catalog No. D21731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21731G 1 g
D21735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21731G Supplier TCI America Supplier No. D21731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2432-12-4
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00134679
InChI Key YXEOEPYIBGTLML-UHFFFAOYSA-N
Synonym 2,6-Dichloro-4-methylphenol
PubChem CID 17077
IUPAC Name 2,6-dichloro-4-methylphenol
SMILES CC1=CC(=C(C(=C1)Cl)O)Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 177.02
Percent Purity ≥96.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,6-Dichloro-p-cresol
EINECSNumber (3)-3660
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.