Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D1750250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1750250G 250 g
D175025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1750250G Supplier TCI America Supplier No. D1750250G

Chemical Identifiers

CAS 2014-83-7
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD00000897
InChI Key LBOBESSDSGODDD-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzyl chloride, 1,3-dichloro-2-chloromethyl benzene, alpha,2,6-trichlorotoluene, 2,6-dichlorobenzylchloride, 1,3-dichloro-2-chloromethyl-benzene, benzene, 1,3-dichloro-2-chloromethyl, toluene, .alpha.,2,6-trichloro, a,2,6-trichlorotoluene, dichlorobenzyl 2,6-chloride, 1,3-ddichloro-2-chloromethyl benzene
PubChem CID 74832
IUPAC Name 1,3-dichloro-2-(chloromethyl)benzene
SMILES C1=CC(=C(C(=C1)Cl)CCl)Cl

Specifications

Melting Point 38°C
Color White-Yellow
Boiling Point 119°C
Quantity 250 g
UN Number 3261
Formula Weight 195.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Dichlorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.