Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloropyridine, 98%

Catalog No. AAA1136422
Click to view available options
Quantity:
100 g
500 g

Catalog No. AAA1136422

Supplier: Thermo Scientific Chemicals A1136422

Only null left
Add to Cart

CAS: 2402-78-0 | C5H3Cl2N | 147.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2402-78-0
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.99
MDL Number MFCD00006244
InChI Key FILKGCRCWDMBKA-UHFFFAOYSA-N
Synonym pyridine, 2,6-dichloro, ccris 1727, 2,6-dichlorpyridine, 2.6-dichloropyridine, pubchem2564, 2,6-dichloro pyridine, 2,6-dichloro-pyridine, acmc-1cbwr, 26dclpy, wln: t6nj bg fg
PubChem CID 16989
IUPAC Name 2,6-dichloropyridine
SMILES ClC1=CC=CC(Cl)=N1

Specifications

Melting Point 84°C to 88°C
Boiling Point 211°C
Odor Unpleasant
Quantity 100 g
UN Number UN2811
Beilstein 108664
Formula Weight 147.99
Percent Purity 98%
Chemical Name or Material 2,6-Dichloropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 219-282-3
RTECSNumber US8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.