Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorostyrene, 98%

Click to view available options
Quantity:
1g
28469-92-3
C8H6Cl2
173.036
MFCD00000579
YJCVRMIJBXTMNR-UHFFFAOYSA-N
2,6-dichlorostyrene,1,3-dichloro-2-vinylbenzene,benzene, 1,3-dichloro-2-ethenyl,2,6-dichlorstyrol,acmc-20ao92,2,6-dichlorostyrene,1,3-dichloro-2-vinylbenzene #,2,6-dichlorostyrene, stabilized 5g
34254
1,3-dichloro-2-ethenylbenzene
C=CC1=C(C=CC=C1Cl)Cl
28469-92-3
C8H6Cl2
173.036
MFCD00000579
YJCVRMIJBXTMNR-UHFFFAOYSA-N
2,6-dichlorostyrene,1,3-dichloro-2-vinylbenzene,benzene, 1,3-dichloro-2-ethenyl,2,6-dichlorstyrol,acmc-20ao92,2,6-dichlorostyrene,1,3-dichloro-2-vinylbenzene #,2,6-dichlorostyrene, stabilized 5g
34254
1,3-dichloro-2-ethenylbenzene
C=CC1=C(C=CC=C1Cl)Cl
Boiling Point 88°C to 90°C (8mmHg)
Quantity 1g
Flash Point 71°C (160°F)
Formula Weight 173.04
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorostyrene

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C