Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diethylaniline, 98%

Catalog No. p-107948
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle

CAS: 579-66-8 | C10H15N | 149.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-66-8
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00007753
InChI Key FOYHNROGBXVLLX-UHFFFAOYSA-N
Synonym benzenamine, 2,6-diethyl, 2,6-diethylbenzenamine, aniline, 2,6-diethyl, 2,6-diethyl aniline, 2-amino-1,3-diethylbenzene, 2,6-diethyl-aniline, ccris 2688, benzamine, 2,6-diethyl, 2,6 diethylaniline, 2,6-diethylphenylamine
PubChem CID 11369
ChEBI CHEBI:27880
IUPAC Name 2,6-diethylaniline
SMILES CCC1=C(C(=CC=C1)CC)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 3°C to 4°C
Color Brown-Red to Yellow
Density 0.9000g/mL
Boiling Point 243°C
Flash Point 123°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5446 to 1.5466
Linear Formula (C2H5)2C6H3NH2
Quantity 250 mL
Beilstein 12,164
Specific Gravity 0.9
Formula Weight 149.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 6-Diethylaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-445-7
RTECSNumber BX3500000
TSCA TSCA
Recommended Storage substance rapidly darkens on exposure to light and air

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.