Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diethynylpyridine 98.0+%, TCI America™
SDP

Catalog No. D50971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50971G 1 g
D5097200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50971G Supplier TCI America Supplier No. D50971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75867-46-8
Molecular Formula C9H5N
Molecular Weight (g/mol) 127.146
MDL Number MFCD09039127
InChI Key VYRLFYTZNXGQIK-UHFFFAOYSA-N
PubChem CID 12579661
IUPAC Name 2,6-diethynylpyridine
SMILES C#CC1=NC(=CC=C1)C#C

Specifications

Melting Point 71°C
Quantity 1 g
Formula Weight 127.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diethynylpyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.