Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,6-Difluoro-3-pyridineboronic Acid (contains varying amounts of Anhydride), TCI America™

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Chemical Name or Material | 2,6-Difluoro-3-pyridineboronic Acid (contains varying amounts of Anhydride) |
CAS | 136466-94-9 |
Color | White-Yellow |
Molecular Formula | C5H4BF2NO2 |
Quantity | 1 g |
MDL Number | MFCD03788245 |
Synonym | 2,6-difluoropyridine-3-boronic acid, 2,6-difluoropyridin-3-yl boronic acid, 2,6-difluoro-3-pyridineboronic acid, 2,6-difluoropyridin-3-yl-3-boronic acid, 2,6-difluoropyridine-3-boronicacid, 2,6-difluoropyridin-3-ylboronicacid, 2,6-difluoro-3-pyridyl boronic acid, 2,6-difluoro-3-pyridylboronic acid, boronic acid, 2,6-difluoro-3-pyridinyl, 2,6-difluoro-3-pyridinyl boronic acid |
InChI Key | LCCZTROJRJFXNV-UHFFFAOYSA-N |
SMILES | B(C1=C(N=C(C=C1)F)F)(O)O |
IUPAC Name | (2,6-difluoropyridin-3-yl)boronic acid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction