Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoro-4-hydroxybenzoic acid, 95%

Catalog No. p-7028728
Click to view available options
Quantity:
250 mg
1 g

CAS: 214917-68-7 | C7H4F2O3 | 174.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214917-68-7
Molecular Formula C7H4F2O3
Molecular Weight (g/mol) 174.10
MDL Number MFCD03094500
InChI Key NFIQGYBXSJQLSR-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-hydroxy-benzoic acid, benzoic acid, 2,6-difluoro-4-hydroxy, pubchem4938, acmc-1cles, ksc495o6f, 4-carboxy-3,5-difluorophenol
PubChem CID 2778776
IUPAC Name 2,6-difluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1F

Specifications

Melting Point 170°C to 171°C (decomposition)
Quantity 250 mg
Formula Weight 174.1
Percent Purity 95%
Chemical Name or Material 2,6-Difluoro-4-hydroxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.