Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH5919503
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218791-32-2
Molecular Formula C14H17BF2O4
Molecular Weight (g/mol) 298.093
MDL Number MFCD12546491
InChI Key CGADBVGBEFYTMK-UHFFFAOYSA-N
Synonym methyl 3,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid, pinacol ester, 2,6-difluoro-4-methoxycarbonyl benzeneboronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenyl boronic acid pinacol ester, 2,6-difluoro-4-methoxycarbonyl phenylboronic acid,pinacol ester, methyl 3,5-difluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, methyl3,5-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate
PubChem CID 46739549
IUPAC Name methyl 3,5-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2F)C(=O)OC)F

Specifications

Quantity 1 g
Formula Weight 298.09
Percent Purity 96%
Chemical Name or Material 2,6-Difluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.