Alfa Aesar™ 2',6'-Difluoroacetophenone, 98%

Manufacturer: Alfa Aesar™ A1022014

Quantity 25g

 View more versions of this product

Catalog No. AAA1022014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13670-99-0
    Molecular Formula C8H6F2O
    Formula Weight 156.13
    MDL Number MFCD00000328
    Beilstein 2046068
    Chemical Name or Material 2',6'-Difluoroacetophenone

    Specifications

    Formula Weight 156.13
    CAS 13670-99-0
    Flash Point 76°C (168°F)
    Boiling Point 76° to 79°C (15mmHg)
    Quantity 25g
    Molecular Formula C8H6F2O
    Beilstein 2046068
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    MDL Number MFCD00000328
    Chemical Name or Material 2',6'-Difluoroacetophenone
    Assay 98%

    Description

    Applications
    2 ft.,6 ft.-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit