Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzeneboronic acid, 98%

Catalog No. p-7026673
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 162101-25-9 | C6H5BF2O2 | 157.911 g/mol

2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162101-25-9
Molecular Formula C6H5BF2O2
Molecular Weight (g/mol) 157.911
MDL Number MFCD00792436
InChI Key DBZAICSEFBVFHL-UHFFFAOYSA-N
Synonym 2,6-difluorobenzeneboronic acid, 2,6-difluorophenyl boronic acid, 2,6-difluorophenyl boranediol, boronic acid, 2,6-difluorophenyl, 2,6-difluorphenylboronic acid, 2,6-difluoro phenylboric acid, boronic acid, b-2,6-difluorophenyl, 2,6-difluorophenylboricacid, pubchem1816, acmc-1celt
PubChem CID 2734336
IUPAC Name (2,6-difluorophenyl)boronic acid
SMILES B(C1=C(C=CC=C1F)F)(O)O

Specifications

Melting Point ∼140°C (decomposition)
Quantity 1 g
Beilstein 8545931
Solubility Information Soluble in methanol.
Formula Weight 157.91
Percent Purity 98%
Chemical Name or Material 2,6-Difluorobenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.