Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzylamine 98.0+%, TCI America™
SDP

Catalog No. D184725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D184725G 25 g
D18475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D184725G Supplier TCI America Supplier No. D184725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69385-30-4
Molecular Formula C7H8F2N
Molecular Weight (g/mol) 144.14
MDL Number MFCD00010144
InChI Key PQCUDKMMPTXMAL-UHFFFAOYSA-O
PubChem CID 123563
IUPAC Name (2,6-difluorophenyl)methanaminium
SMILES [NH3+]CC1=C(F)C=CC=C1F

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 g
UN Number 2735
Synonym 2,6-difluorobenzylamine, 2,6-difluorophenyl methanamine, 2,6-difluorobenzyl amine, benzenemethanamine, 2,6-difluoro, 1-2,6-difluorophenyl methanamine, 2,6-difluorophenyl methylamine, 2, 6-difluorobenzylamine, pubchem2235, 2,6-difluoro-benzylamine, acmc-1b2iy
Formula Weight 143.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.