Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoronitrobenzene, 98%, Thermo Scientific™

Catalog No. p-3770926
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 19064-24-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00192035
InChI Key SSNCMIDZGFCTST-UHFFFAOYSA-N
Synonym 2,6-difluoronitrobenzene, 1,3-difluoro-2-nitro-benzene, benzene, 1,3-difluoro-2-nitro, 2,6-difluoro nitrobenzene, pubchem4403, 1,3-difluoronitrobenzene, acmc-1buv1, 2,6-difluoro-nitrobenzene, 2,6-difluoro nitro benzene, 1,3-difluoro-2-nitrobenzen
PubChem CID 87922
IUPAC Name 1,3-difluoro-2-nitrobenzene
SMILES C1=CC(=C(C(=C1)F)[N+](=O)[O-])F

Specifications

Density 1.5030g/mL
Boiling Point 91.0°C to 92.0°C (11.0 mmHg)
Flash Point 88°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula F2C6H3NO2
Quantity 5 g
Specific Gravity 1.503
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 2, 6-Difluoronitrobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 242-793-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.