Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorophenylacetic acid, 99%, Thermo Scientific™

Catalog No. p-110901
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-28-6
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00010001
InChI Key FUGDCKXBUZFEON-UHFFFAOYSA-N
Synonym 2,6-difluorophenylacetic acid, 2-2,6-difluorophenyl acetic acid, 2,6-difluorophenyl acetic acid, 2,6-difluoro-benzene acetic acid, benzeneacetic acid, 2,6-difluoro, 2,6-difluorophenylaceticacid, pubchem8397, acmc-209xw7, rarechem al bo 0249
PubChem CID 123585
IUPAC Name 2-(2,6-difluorophenyl)acetic acid
SMILES C1=CC(=C(C(=C1)F)CC(=O)O)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 98°C to 101°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula F2C6H3CH2COOH
Quantity 1 g
Formula Weight 172.13
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Difluorophenylacetic acid, 99%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 285-289-3
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.