Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diisopropylaniline, 90+%

Catalog No. p-7045447
Click to view available options
Quantity:
100 g
500 g

CAS: 24544-04-5 | C12H19N | 177.291 g/mol

2,6-Diisopropylaniline acts as an intermediate used in the production of carbodiimides stabilizers, synthetic resins, antioxidants and active pharmaceutical ingredients. It is also used in the preparation of multitopic Schiff-base ligand precursors and 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylthioxanthene. It also undergoes condensation reaction with triacetylmethane to get -[1-(2,6-diisopropylphenylamino)ethylidene]pentane-2,4-dione. Further, it is used to prepare N-heterocyclic carbene complexes for alfa-arylation of acyclic ketones, amination of haloarenes and aqueous Suzuki coupling. In addition to this, it is used in the prepartion of organocatalyst based on naphthalene diimides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diisopropylaniline acts as an intermediate used in the production of carbodiimides stabilizers, synthetic resins, antioxidants and active pharmaceutical ingredients. It is also used in the preparation of multitopic Schiff-base ligand precursors and 4,5-bis(2,6-diisopropylanilino)-2,7-di-tert-butyl-9,9-dimethylthioxanthene. It also undergoes condensation reaction with triacetylmethane to get -[1-(2,6-diisopropylphenylamino)ethylidene]pentane-2,4-dione. Further, it is used to prepare N-heterocyclic carbene complexes for alfa-arylation of acyclic ketones, amination of haloarenes and aqueous Suzuki coupling. In addition to this, it is used in the prepartion of organocatalyst based on naphthalene diimides.

Solubility
Miscible with ether and alcohol. Slightly miscible with water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24544-04-5
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00008887
InChI Key WKBALTUBRZPIPZ-UHFFFAOYSA-N
Synonym 2,6-diisopropylaniline, benzenamine, 2,6-bis 1-methylethyl, 2,6-diisopropyl aniline, aniline, 2,6-diisopropyl, diisopropylaniline, 2,6-diisopropylphenylamine, 2,6-bis propan-2-yl aniline, 2,6-dilsopropylaniline, 2,6-di propan-2-yl aniline, 2,6-bis 1-methylethyl aniline
PubChem CID 32484
IUPAC Name 2,6-di(propan-2-yl)aniline
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N

Specifications

Melting Point -54°C
Density 0.94
Boiling Point 254°C to 257°C
Flash Point 125°C (257°F)
Refractive Index 1.533
Quantity 100 g
Beilstein 2208763
Solubility Information Miscible with ether and alcohol. Slightly miscible with water.
Formula Weight 177.29
Percent Purity ≥90%
Chemical Name or Material 2,6-Diisopropylaniline
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 246-305-4
RTECSNumber BX4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.