Learn More
2,6-Dimethoxyaniline, 97%, Thermo Scientific Chemicals
Description
2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,6-Dimethylaniline degradation by Fenton process has been studied in depth for the purpose of learning more about the reactions involved in the oxidation of 2,6-dimethylaniline under various reaction conditions. Using a previously developed gas chromatographic-mass spectrometric assay, hemoglobin adducts of 2,6-dimethylaniline were detected covalently bound to rat hemoglobin after administration of either 2,6-dimethylaniline or lidocaine.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 76°C to 78°C |
Odor | Characteristic |
Quantity | 5 g |
UN Number | UN2811 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 153.18 |
Percent Purity | 97% |
Chemical Name or Material | 2,6-Dimethoxyaniline |
Safety and Handling
Hazard Category | H302+H332-H311-H315-H319-H335-H373 |
Hazard Statement | GHS H Statement H311-H373-H302-H332 Toxic in contact with skin. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.