Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,6-Dimethoxybenzaldehyde, 98+%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.
Solubility
Soluble in Methanol (almost transparency).
Notes
Air sensitive. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, air.
2,6-Dimethoxybenzaldehyde has been used in the preparation of 2,6-dihydroxybenzaldehyde by demethylation with AlBr3.
Solubility
Soluble in Methanol (almost transparency).
Notes
Air sensitive. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent, air.

Specifications
Specifications
Melting Point | 96°C to 98°C |
Boiling Point | 285°C |
Quantity | 5 g |
Beilstein | 908138 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in Methanol (almost transparency). |
Formula Weight | 166.18 |
Percent Purity | 99% |
Chemical Name or Material | 2,6-Dimethoxybenzaldehyde |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction