Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxybenzeneboronic acid, 97+%

Catalog No. AAB2430506
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 23112-96-1 | C8H11BO4 | 181.982 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23112-96-1
Molecular Formula C8H11BO4
Molecular Weight (g/mol) 181.982
MDL Number MFCD01318987
InChI Key BKWVXPCYDRURMK-UHFFFAOYSA-N
Synonym 2,6-dimethoxyphenyl boronic acid, 2,6-dimethoxyphenylboronicacid, 2,6-dimethoxybenzeneboronic acid, 2,6-dimethoxyphenyl-boronic acid, 2,6-dimethoxyphenyl boranediol, boronic acid, 2,6-dimethoxyphenyl, pubchem1825, acmc-1clhv, ksc201q0f, 2,6-dimethoxy-phenylboronic acid
PubChem CID 2734343
IUPAC Name (2,6-dimethoxyphenyl)boronic acid
SMILES B(C1=C(C=CC=C1OC)OC)(O)O

Specifications

Melting Point 106°C to 108°C
Quantity 5 g
Beilstein 3032352
Formula Weight 181.98
Percent Purity ≥97%
Chemical Name or Material 2,6-Dimethoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.