Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxyphenol, 99%

Catalog No. AAB2425322
Click to view available options
Quantity:
25 g
100 g

CAS: 91-10-1 | C8H10O3 | 154.17 g/mol

It is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-10-1
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00064434
InChI Key KLIDCXVFHGNTTM-UHFFFAOYSA-N
Synonym syringol, pyrogallol 1,3-dimethyl ether, 1,3-dimethyl pyrogallate, phenol, 2,6-dimethoxy, 2-hydroxy-1,3-dimethoxybenzene, 1,3-di-o-methylpyrogallol, 1,3-dimethoxy-2-hydroxybenzene, 2,6-dimethoxy-phenol, pyrogallol dimethylether, 2,6-dwumetoksyfenol
PubChem CID 7041
ChEBI CHEBI:955
IUPAC Name 2,6-dimethoxyphenol
SMILES COC1=CC=CC(OC)=C1O

Specifications

Melting Point 50°C to 57°C
Boiling Point 261°C
Flash Point 140°C (284°F)
Quantity 100 g
Beilstein 1526871
Solubility Information Soluble in water.
Formula Weight 154.16
Percent Purity 99%
Chemical Name or Material 2,6-Dimethoxyphenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-041-1
RTECSNumber SL0900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.