Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxyphenol, 99%

Catalog No. p-114400
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 91-10-1 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-10-1
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00064434
InChI Key KLIDCXVFHGNTTM-UHFFFAOYSA-N
Synonym syringol, pyrogallol 1,3-dimethyl ether, 1,3-dimethyl pyrogallate, phenol, 2,6-dimethoxy, 2-hydroxy-1,3-dimethoxybenzene, 1,3-di-o-methylpyrogallol, 1,3-dimethoxy-2-hydroxybenzene, 2,6-dimethoxy-phenol, pyrogallol dimethylether, 2,6-dwumetoksyfenol
PubChem CID 7041
ChEBI CHEBI:955
IUPAC Name 2,6-dimethoxyphenol
SMILES COC1=CC=CC(OC)=C1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 50°C to 56°C
Color Brown to Gray or White
Boiling Point 261°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3OH
Quantity 25 g
Beilstein 06,1081
Solubility Information Solubility in water: 2% (13°C). Other solubilities: soluble in ethyl ether,ethanol
Formula Weight 154.17
Percent Purity 99%
Physical Form Crystalline Powder, Crystals, or Crystalline Solid
Chemical Name or Material 2, 6-Dimethoxyphenol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-041-1
RTECSNumber SL0900000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.