Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxytoluene, 98%

Catalog No. aab2260622
Click to view available options
Quantity:
100 g

CAS: 5673-07-4 | C9H12O2 | 152.19 g/mol

2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethoxytoluene is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5673-07-4

Specifications

Melting Point 37°C to 41°C
Boiling Point 222°C
Flash Point 96°C (204°F)
Odor Aromatic
Molecular Formula C9H12O2
Quantity 100 g
MDL Number MFCD00008374
Beilstein 2045327
Synonym 2,6-dimethoxytoluene, benzene, 1,3-dimethoxy-2-methyl, 2-methyl-1,3-bis methyloxy benzene, 1,3-dimethoxy-2-methylbenzol, pubchem4545, acmc-1aqu0, 3-chloro-6-hydroxybiphenyl, 1,3-dimethoxy-2-methyl-benzene
Solubility Information Insoluble in water.
InChI Key FPEUDBGJAVKAEE-UHFFFAOYSA-N
SMILES CC1=C(C=CC=C1OC)OC
IUPAC Name 1,3-dimethoxy-2-methylbenzene
Molecular Weight (g/mol) 152.19
PubChem CID 79755
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2,6-Dimethoxytoluene
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-131-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.