Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-4-heptanone, technical, remainder mainly 4,6-dimethyl-2-heptanone

Catalog No. p-118117
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Metal drum

CAS: 108-83-8 | C9H18O | 142.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-83-8, 19549-80-5
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00008940
InChI Key PTTPXKJBFFKCEK-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-heptanone, diisobutyl ketone, isovalerone, isobutyl ketone, valerone, diisobutylketone, diisobutilchetone, s-diisopropylacetone, di-isobutylcetone, dibk
PubChem CID 7958
IUPAC Name 2,6-dimethylheptan-4-one
SMILES CC(C)CC(=O)CC(C)C

Specifications

CAS Min % 75.0
CAS Max % 100.0
Melting Point -46°C
Color Colorless to Yellow
Density 0.8090g/mL
Boiling Point 169°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 80%
Packaging Glass Bottle
Refractive Index 1.4120 to 1.415
Linear Formula (CH3)2CHCH2COCH2CH(CH3)2
Quantity 1 L
Beilstein 01, 710
Specific Gravity 0.809
Solubility Information Solubility in water: slightly soluble (0.05g/100 mL). Other solubilities: miscible with most organic solvents
Viscosity 1.05 mPa.s (20°C)
Formula Weight 142.24
Percent Purity ≥75% (typically) exact percentage on the certificate of analysis, 96% min. (main compound plus isomer)
Grade Technical
Physical Form Liquid
Chemical Name or Material 2, 6-Dimethyl-4-heptanone, tech., 80%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 203-620-1
RTECSNumber MJ5775000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.