Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-4-pyrone, 99%

Catalog No. p-7020739
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 1004-36-0 | C7H8O2 | 124.139 g/mol

2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressure.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1004-36-0
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00006579
InChI Key VSYFZULSKMFUJJ-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4h-pyran-4-one, 2,6-dimethyl-4-pyrone, 2,6-dimethyl-gamma-pyrone, 2,6-dimethyl-4-pyranone, 4h-pyran-4-one, 2,6-dimethyl, gamma-2,6-dimethylpyrone, unii-v2d262mf0l, 2,6-dimethyl-pyran-4-one, 2,6-dimethyl-.gamma.-pyrone, ccris 3601
PubChem CID 13862
IUPAC Name 2,6-dimethylpyran-4-one
SMILES CC1=CC(=O)C=C(O1)C

Specifications

Melting Point 132°C to 136°C
Boiling Point 248°C to 250°C
Quantity 10 g
Beilstein 107418
Solubility Information Slightly soluble in water.
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-4-pyrone
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-719-1
RTECSNumber UQ0780000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.