Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylbenzonitrile 98.0+%, TCI America™
SDP

Catalog No. D492725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D492725G 25 g
D49275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D492725G Supplier TCI America Supplier No. D492725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6575-13-9
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00037166
InChI Key QSACPWSIIRFHHR-UHFFFAOYSA-N
Synonym benzonitrile, 2,6-dimethyl, unii-1zh37s5b0z, 2,6-dimethylbenzenecarbonitrile, 2,6-dimethyl-benzonitrile, 2-cyano-m-xylene, ksc494c5t, 2,6-dimethylbenz0nitrile, 3-cyano-2,4-dimethylphenyl
PubChem CID 81057
IUPAC Name 2,6-dimethylbenzonitrile
SMILES CC1=C(C(=CC=C1)C)C#N

Specifications

Melting Point 90°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 93°C
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,6-Dimethylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.