Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylhydroquinone 98.0+%, TCI America™
SDP

Catalog No. D266725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D266725G 25 g
D26675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D266725G Supplier TCI America Supplier No. D266725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 654-42-2
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00016466
InChI Key SGWZVZZVXOJRAQ-UHFFFAOYSA-N
Synonym 2,5-Dihydroxy-m-xylene
PubChem CID 69560
IUPAC Name 2,6-dimethylbenzene-1,4-diol
SMILES CC1=CC(O)=CC(C)=C1O

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 138.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethylhydroquinone
EINECSNumber (4)-0057
RTECSNumber MX5900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.