Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylphenyl Isocyanate 98.0+%, TCI America™
SDP

Catalog No. I041025G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 28556-81-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00002012
InChI Key YQLRKXVEALTVCZ-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl isocyanate, benzene, 2-isocyanato-1,3-dimethyl, 2,6-dimethylphenylisocyanate, 2,6-xylyl isocyanate, unii-2ql5mu0e0c, 2ql5mu0e0c, isocyanic acid 2,6-dimethylphenyl ester, 2,6-dimethylbenzenisocyanate, 2,6-dimethyl phenyl isocyanate, 2-isocyanato-1,3-dimethyl-benzene
PubChem CID 98787
IUPAC Name 2-isocyanato-1,3-dimethylbenzene
SMILES CC1=CC=CC(C)=C1N=C=O

Specifications

Color Yellow
Boiling Point 113°C
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 2206
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,6-Dimethylphenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.