Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylphenyl isothiocyanate, 98%

Catalog No. AAA1948014
Click to view available options
Quantity:
25 g

CAS: 19241-16-8 | C9H9NS | 163.238 g/mol

2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylphenyl isocyanate has been used in the preparation of derivatized β-cyclodextrins, used as chiral stationary phase in normal-phase liquid chromatography, tris( 2,6-dimethylphenylimido)methylrhenium (VII), 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19241-16-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00038715
InChI Key UULUECCNPPJFBU-UHFFFAOYSA-N
PubChem CID 87976
IUPAC Name 2-isothiocyanato-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)N=C=S

Specifications

Density 1.085
Boiling Point 247°C
Quantity 25 g
Odor Pungent
Refractive Index 1.625
UN Number UN2922
Beilstein 637195
Synonym 2,6-dimethylphenyl isothiocyanate, 2,6-xylyl isothiocyanate, 2,6-dimethylphenylisothiocyanate, benzene, 2-isothiocyanato-1,3-dimethyl, 2-isothiocyanato-1,3-dimethyl-benzene, benzene,2-isothiocyanato-1,3-dimethyl, 2,6-dimethylbenzenisothiocyanate, acmc-20ao7z, 2,6-dimethylphenyl-isothiocyanate, 2,6-dimethylphenyl-1-isothiocyanate
Solubility Information Hydrolyzes in water.
Formula Weight 163.24
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylphenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 242-906-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.