Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,6-Dimethylpyrazine, 99%
Click to view available options
Quantity:
25 g
Description
2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.

Specifications
Specifications
Melting Point | 38°C to 43°C |
Boiling Point | 153°C to 154°C |
Flash Point | 52°C (125°F) |
Odor | Aromatic |
Refractive Index | 1.5 |
Quantity | 25 g |
UN Number | UN1325 |
Beilstein | 1726 |
Solubility Information | Soluble in water. |
Formula Weight | 108.14 |
Show More |
Safety and Handling
Hazard Category | H228-H302 |
Hazard Statement | GHS H Statement H228-H302-H315-H319-H335 Flammable solid. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P240-P241-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c |
DOTInformation | Hazard Class: 4.1; Packaging Group: II |
EINECSNumber | 203-589-4 |
RTECSNumber | UQ2975000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction