Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylpyridinium p-Toluenesulfonate, TCI America™
SDP

Catalog No. d177125g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 93471-41-1
Molecular Formula C14H17NO3S
Molecular Weight (g/mol) 279.354
MDL Number MFCD00060191
InChI Key ZXFCGHQVBSUJLX-UHFFFAOYSA-N
Synonym 2,6-Litidinium p-Toluenesulfonate
PubChem CID 10880460
IUPAC Name 2,6-dimethylpyridin-1-ium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].CC1=[NH+]C(=CC=C1)C

Specifications

Melting Point 117°C
Quantity 25 g
Formula Weight 279.35
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethylpyridinium p-Toluenesulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.