Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylquinoline 99.0+%, TCI America™
SDP

Catalog No. D123625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D123625G 25 g
D12361G 1 g
D12365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D123625G Supplier TCI America Supplier No. D123625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 877-43-0
Molecular Formula C11H11N
Molecular Weight (g/mol) 157.216
MDL Number MFCD00006762
InChI Key JJPSZKIOGBRMHK-UHFFFAOYSA-N
Synonym p-toluquinaldine, quinoline, 2,6-dimethyl, 6-methylquinaldine, unii-kst0m1t4mb, 2,6-dimethyl-quinoline, kst0m1t4mb, pubchem5876, quinoline,6-dimethyl, acmc-209qpt, 2,6-dimethylquinoline;
PubChem CID 13414
IUPAC Name 2,6-dimethylquinoline
SMILES CC1=CC2=C(C=C1)N=C(C=C2)C

Specifications

Melting Point 58°C
Color White
Boiling Point 267°C
Quantity 25 g
Formula Weight 157.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.