Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diphenylphenol 98.0+%, TCI America™
SDP

Catalog No. D256325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D256325G 25 g
D25635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D256325G Supplier TCI America Supplier No. D256325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2432-11-3
Molecular Formula C18H14O
Molecular Weight (g/mol) 246.31
MDL Number MFCD00009716
InChI Key ATGFTMUSEPZNJD-UHFFFAOYSA-N
Synonym 1,1':3',1-terphenyl-2'-ol, m-terphenyl-2'-ol, 2,6-diphenyl phenol, 2'-hydroxy-m-terphenyl, unii-la93w35m96, 3-phenyl-1,1'-biphenyl-2-ol, 2,6-di-phenylphenol, 2,6,-diphenylphenol, 2,6-di phenyl phenol
PubChem CID 75512
IUPAC Name 2,6-diphenylphenol
SMILES C1=CC=C(C=C1)C2=C(C(=CC=C2)C3=CC=CC=C3)O

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 25 g
Formula Weight 246.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diphenylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.