Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diphenylpyridine 99.0+%, TCI America™
SDP

Catalog No. D192225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D192225G 25 g
D19225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D192225G Supplier TCI America Supplier No. D192225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3558-69-8
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
MDL Number MFCD00006284
InChI Key PJUOHDQXFNPPRF-UHFFFAOYSA-N
Synonym pyridine, 2,6-diphenyl, unii-zi75v15y1t, 2,6-diphenyl-pyridine, 2,6-diphenyl pyridine, maybridge3_000511, acmc-1cq41, pjuohdqxfnpprf-uhfffaoysa
PubChem CID 72920
IUPAC Name 2,6-diphenylpyridine
SMILES C1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=CC=C3

Specifications

Color White-Yellow
Boiling Point 398°C
Quantity 25 g
Formula Weight 231.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diphenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.