Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Di(1-pyrenyl)-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
SDP

Catalog No. D49321G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49321G 1 g
D4932200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49321G Supplier TCI America Supplier No. D49321G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 886456-80-0
Molecular Formula C57H32
Molecular Weight (g/mol) 716.883
MDL Number MFCD12022456
InChI Key MJXRNMQMLMXICM-UHFFFAOYSA-N
PubChem CID 86242657
IUPAC Name 2',7'-di(pyren-1-yl)-9,9'-spirobi[fluorene]
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=C(C=CC(=C5)C6=C7C=CC8=CC=CC9=C8C7=C(C=C9)C=C6)C1=C4C=C(C=C1)C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1

Specifications

Quantity 1 g
Formula Weight 716.88
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Di(1-pyrenyl)-9,9′-spirobi[9H-fluorene]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.