Learn More
2,7-Diaminofluorene, 97%
Description
It is used as substitute for the carcinogenic benzidine in the chromogenic assay of peroxidases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as substitute for the carcinogenic benzidine in the chromogenic assay of peroxidases.
Solubility
Slightly soluble in cold water. Soluble in hot water. Sparingly soluble in ethanol. and acetone. Insoluble in ether.
Notes
Store at 4°C. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 160°C to 162°C |
| Quantity | 5 g |
| Beilstein | 2099859 |
| Merck Index | 14,4156 |
| Solubility Information | Slightly soluble in cold water. Soluble in hot water. Sparingly soluble in ethanol. and acetone. Insoluble in ether. |
| Formula Weight | 196.25 |
| Percent Purity | ≥97% |
| Chemical Name or Material | 2,7-Diaminofluorene |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 208-377-5 |
| RTECSNumber | LL6980000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.