Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromonaphthalene, 99%

Catalog No. p-7037376
Click to view available options
Quantity:
1 g
5 g

CAS: 58556-75-5 | C10H6Br2 | 285.97 g/mol

It is an intermediate in dyestuffs and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate in dyestuffs and pharmaceuticals.

Solubility
Soluble in acetonitrile.

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58556-75-5
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD00238580
InChI Key ODJZWBLNJKNOJK-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dibromo, acmc-209m5e, ksc490o3j, 2,7-dibromonaphthalene hplc
PubChem CID 625355
IUPAC Name 2,7-dibromonaphthalene
SMILES BrC1=CC2=CC(Br)=CC=C2C=C1

Specifications

Melting Point 139°C to 142°C
Quantity 1 g
UN Number UN3077
Beilstein 1862041
Solubility Information Soluble in acetonitrile.
Formula Weight 285.97
Percent Purity 99%
Chemical Name or Material 2,7-Dibromonaphthalene
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.