Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',7'-Dichlorofluorescein, ACS reagent

Catalog No. p-4440196
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle

CAS: 76-54-0 | C20H10Cl2O5 | 401.195 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-54-0
Molecular Formula C20H10Cl2O5
Molecular Weight (g/mol) 401.195
InChI Key VFNKZQNIXUFLBC-UHFFFAOYSA-N
Synonym 2',7'-Dichloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one
PubChem CID 64944
ChEBI CHEBI:51596
IUPAC Name 2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl

Specifications

Formula Weight 401.2
Grade ACS Reagent
Packaging Glass bottle
Density 0.7900g/mL
Quantity 1 g
Chemical Name or Material 2', 7'-Dichlorofluorescein

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.