Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',7'-Dichlorofluorescein 95.0+%, TCI America™
SDP

Catalog No. D037125G
Click to view available options
Quantity:
1 g
25 g

Chemical Identifiers

CAS 76-54-0
Molecular Formula C20H10Cl2O5
Molecular Weight (g/mol) 401.195
MDL Number MFCD00005047
InChI Key VFNKZQNIXUFLBC-UHFFFAOYSA-N
Synonym 2',7'-dichlorofluorescein, dichlorofluorescein, unii-56nqm5uzt1, 2,7-dichlorofluorescein, 56nqm5uzt1, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxy, 2',7'-dichloro-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxyspiro 2-benzofuran-3,9'-xanthene-1-one, 11,16-dichloro-12,15-dihydroxyspiro 3-hydroisobenzofuran-3,9'-xanthene-1-one
PubChem CID 64944
ChEBI CHEBI:51596
IUPAC Name 2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl

Specifications

Melting Point 280°C
Quantity 25 g
Formula Weight 401.20
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,7′-Dichlorofluorescein [for Fluorescent indicator]
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.