Alfa Aesar™ 2,7-Dihydroxynaphthalene, 97%

Manufacturer: Alfa Aesar™ A1022930

Quantity 250g

 View more versions of this product

Catalog No. AAA1022930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 582-17-2
    Molecular Formula C10H8O2
    Formula Weight 160.17
    MDL Number MFCD00004085
    Beilstein 2042383
    Synonym 2,7-Naphthalenediol
    Chemical Name or Material 2,7-Dihydroxynaphthalene

    Specifications

    Formula Weight 160.17
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C10H8O2
    CAS 582-17-2
    TSCA TSCA
    Beilstein 2042383
    RTECS Number QJ4780000
    MDL Number MFCD00004085
    Synonym 2,7-Naphthalenediol
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 250g
    Health Hazard 1 Warning
    Melting Point 183° to 188°C
    Chemical Name or Material 2,7-Dihydroxynaphthalene
    Assay 97%

    Description

    Applications
    2,7-Naphthalenediol is a reagent used in the preparation of monomers of high carbon materials. Also used in the synthesis of splitomicin analogues. Used as Starting material for the synthesis of sulfonic acids and divinylnaphthalenes. It is also used in making dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.

    Solubility
    Soluble in water.

    Notes
    Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit