Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 2-8 CELLsium™ Medium for Short-Term Cell Storage

Catalog No. MP092780354 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mL
500 mL

2-8 CELLsius is a cytoprotective, protein free, ready to use bio-solution without DMSO for the short-term storage of cells or tissue.

  • For short-term storage, add cold (2 to 8 °C) 2-8 CELLsium at a recommended cell concentration of 1 x 106 mL to 8 x 106 mL
  • The 2-8 CELLsium can be removed after centrifugation, cell concentration adjusted and transferred to an appropriate culture flask with growth medium
  • It will keep the cells healthy while in a temporary senescent phase, unlike regular growth media, which cannot
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Sterility Sterile
For Use With (Application) Short-term cell storage
Quantity 500 mL
Content And Storage Stored at 2 to 8°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.