Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,9-Diacetylguanine, 96%, Thermo Scientific™

Catalog No. AC459211000
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 3056-33-5
Molecular Formula C9H9N5O3
Molecular Weight (g/mol) 235.20
MDL Number MFCD00142116
InChI Key GILZZWCROUGLIS-UHFFFAOYSA-N
PubChem CID 76461
IUPAC Name N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide
SMILES CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O

Specifications

Melting Point 285.0°C
Infrared Spectrum Authentic
Quantity 100 g
Synonym n2,9-diacetylguanine, n,9-diacetylguanine, n 2 ,9-diacetylguanine, 2,9-diacetylguanine, 2,9-diacetateguanine, n-9-acetyl-6-oxo-6,9-dihydro-1h-purin-2-yl acetamide, unii-vuw2b9yr95, 2,9-diacetyl guanine, vuw2b9yr95, n-9-acetyl-6-oxo-3h-purin-2-yl acetamide
Formula Weight 235.2
Percent Purity 96%
Chemical Name or Material 2,9-Diacetylguanine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 221-287-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.