Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol 98.0+%, TCI America™
SDP

Catalog No. F091225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F091225G 25 g
F09125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F091225G Supplier TCI America Supplier No. F091225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105496-31-9
Molecular Formula C17H17NO3
Molecular Weight (g/mol) 283.33
MDL Number MFCD00235927
InChI Key XLIFWDZVNRWYKV-UHFFFAOYSA-N
Synonym 2-(Fmoc-amino)-1-ethanol, N-Fmoc-ethanolamine
PubChem CID 2817739
IUPAC Name (9H-fluoren-9-yl)methyl N-(2-hydroxyethyl)carbamate
SMILES OCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 141°C
Color White
Quantity 25 g
Formula Weight 283.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-1-ethanol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.