Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamido-6-aminopyridine 98.0+%, TCI America™
SDP

Catalog No. A270725G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1075-62-3
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.17
MDL Number MFCD01099052
InChI Key CCVCJYWIEHEXOR-UHFFFAOYSA-N
PubChem CID 3767345
IUPAC Name N-(6-aminopyridin-2-yl)acetamide
SMILES CC(=O)NC1=CC=CC(N)=N1

Specifications

Melting Point 160°C
Color White
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-6-aminopyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.