Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamidobenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. A29261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29261G 1 g
A2926200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29261G Supplier TCI America Supplier No. A29261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13493-47-5
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD06739581
InChI Key OWMJAQBUFVTERI-UHFFFAOYSA-N
PubChem CID 326664
IUPAC Name N-(2-formylphenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1C=O

Specifications

Melting Point 67°C
Color Yellow
Boiling Point 103°C
Quantity 1 g
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamidobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.